missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum wire, 99.95% (metals basis), Alfa Aesar™

Form: approx. 3.26g/m; Temper: soft

Manufacturer:  Alfa Aesar 10349G5

 View more versions of this product

Catalog No. AA10349G5



Specifications

Specifications

Tantalum
Annealed
Gray to silver wire
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
Odorless
23956
Wire, 0.5mm (0.02 in.) dia.
5m
7440-25-7
99.95% (Metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
180.948
CHEBI:33348
Gray to Silver

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

EINECSNumber : 231-135-5

Documents