Learn More
Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)
CAS: 20219-33-4 | C13H27O6Ta | 460.30 g/mol
Supplier: Thermo Scientific Chemicals 03963004
Description
Tantalum(V) tetraethoxide 2,4-pentanedionate is used in pharmaceutical, biochemical and chemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tantalum(V) tetraethoxide 2,4-pentanedionate | |
| 45°C | |
| 95°C (0.5 mmHg) | |
| (metals basis) | |
| MFCD00145349 | |
| Moisture sensitive | |
| Reacts with water. | |
| [Ta+5].CC[O-].CC[O-].CC[O-].CC[O-].C\C([O-])=C\C(C)=O | |
| 460.30 | |
| 460.3 | |
| Solid |
| 20219-33-4 | |
| 1.5 g/mL | |
| >65°C (149°F) | |
| C13H27O6Ta | |
| 2 g | |
| tantalum v tetraethoxide 2,4-pentanedionate | |
| LSALXRIVOAYYJW-GLUPTGNJSA-M | |
| ethanol;(E)-4-hydroxypent-3-en-2-one;tantalum | |
| 122130355 | |
| 99.99% |
Chemical Identifiers
| 20219-33-4 | |
| 460.30 | |
| LSALXRIVOAYYJW-GLUPTGNJSA-M | |
| 122130355 |
| C13H27O6Ta | |
| MFCD00145349 | |
| tantalum v tetraethoxide 2,4-pentanedionate | |
| [Ta+5].CC[O-].CC[O-].CC[O-].CC[O-].C\C([O-])=C\C(C)=O |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only