missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum(V) oxide, Spectrographic Grade, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 7311822

 View more versions of this product

Catalog No. AA7311822



Specifications

Specifications

Tantalum (V) oxide
(metals basis)
MFCD00011254
441.89
99.5%
8.2g/mL
1800°C
1314-61-0
Ta2O5
14,9056
Powder
Sprectrographic
Odorless
100g

Chemical Identifiers

1314-61-0
Ta2O5
MFCD00011254
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents