Please login to your online account to display your discounted pricing

Tantalum(V) oxide, 99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tantalum(V) oxide
CAS 1314-61-0
Molecular Formula Ta2O5
Formula Weight 441.89
MDL Number MFCD00011254
Merck Index 14,9056
Synonym Tantalum pentoxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1470930

 
alfa aesar
1470930
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 441.89
Quantity 250g
Chemical Name or Material Tantalum(V) oxide
TSCA TSCA
Odor Odorless
Physical Form Powder
Assay 99%
Assay Percent Notes (metals basis)
Molecular Formula Ta2O5
MDL Number MFCD00011254
CAS 1314-61-0
Merck Index 14,9056
Synonym Tantalum pentoxide
Melting Point 1800°C
Solubility Information Insoluble in water, alcohol, mineral acids. Soluble in HF
Density 8.2