Please login to your online account to display your discounted pricing

Tantalum(V) oxide, 99% (metal basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tantalum(V) oxide
CAS 1314-61-0
Physical Form Powder
Assay Percent Range 99% (metals basis)
Molecular Formula Ta2O5
Formula Weight 441.89
MDL Number MFCD00011254
Synonym Tantalum pentoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1470909

  SDS
alfa aesar
1470909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1470918

  SDS
alfa aesar
1470918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 441.89
Quantity 10g
Physical Form Powder
EINECS Number 215-238-2
CAS 1314-61-0
Synonym Tantalum pentoxide
Assay Percent Range 99% (metals basis)
Molecular Formula Ta2O5
Chemical Name or Material Tantalum(V) oxide
MDL Number MFCD00011254