Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum(V) oxide, 99.95% (metals basis excluding Nb), Nb <200ppm

Chemical Identifiers

Chemical Name or Material Tantalum(V) oxide
CAS 1314-61-0
Formula Weight 441.89
MDL Number MFCD00011254
Merck Index 14,9056
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3634314

  SDS
alfa aesar™
3634314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3634322

  SDS
alfa aesar™
3634322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tantalum(V) oxide
CAS 1314-61-0
TSCA TSCA
Physical Form Sintered Lump
Assay 99.95%
Assay Percent Notes (metals basis excluding Nb), Nb <200ppm
Formula Weight 441.89
MDL Number MFCD00011254
Merck Index 14,9056
Density 8.2g/mL
Odor Odorless
Particle Size 3 to 12mm
Melting Point 1800°C
Quantity 25g