Please login to your online account to display your discounted pricing

Tantalum(V) oxide, 99.85% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tantalum(V) oxide
CAS 1314-61-0
Physical Form -325 Mesh Powder
Assay Percent Range 99.85% (metals basis)
Molecular Formula Ta2O5
Formula Weight 441.89
MDL Number MFCD00011254
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1133614

  SDS
alfa aesar
1133614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1133622

  SDS
alfa aesar
1133622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1133636

  SDS
alfa aesar
1133636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Tantalum(V) oxide
EINECS Number 215-238-2
Physical Form -325 Mesh Powder
Assay Percent Range 99.85% (metals basis)
Molecular Formula Ta2O5
Formula Weight 441.89
MDL Number MFCD00011254
CAS 1314-61-0
Density 8.2g/mL