missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum(V) Fluoride, 99.9% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1405218

 View more versions of this product

Catalog No. AA1405218



Specifications

Specifications

Tantalum(V) Fluoride
99.9%, metal basis
MFCD00042542
YRGLXIVYESZPLQ-UHFFFAOYSA-I
pentafluorotantalum
82218
Powder
Moisture sensitive
50g
7783-71-3
F5Ta
tantalum pentafluoride, tantalum v fluoride, tantalum fluoride, tantalum fluoride taf5, taf5, tantalum v fluoride, tantalum fluoride taf5, tantalum v fluoride
F[Ta](F)(F)(F)F
275.94
275.94
4.74
Decomposes in water forming tantalum oxyfluoride

Chemical Identifiers

7783-71-3
275.94
YRGLXIVYESZPLQ-UHFFFAOYSA-I
82218
F[Ta](F)(F)(F)F
F5Ta
MFCD00042542
tantalum pentafluoride, tantalum v fluoride, tantalum fluoride, tantalum fluoride taf5, taf5, tantalum v fluoride, tantalum fluoride taf5, tantalum v fluoride
pentafluorotantalum
Safety and Handling

Safety and Handling

Harmful by inhalation, in contact with skin and if swallowed. Causes burns.

DOTInformation : DOT Class: 8

EINECSNumber : 232-022-3

Documents