missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum(V) ethoxide, optical grade, 99.95% (metal basis)

Manufacturer:  Alfa Aesar™ L1603306

 View more versions of this product

Catalog No. AAL1603306

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Tantalum(V) ethoxide
Optical Grade
C10H25O5Ta
406.25
UN2924
Tantalum(V) ethylate
18° to 19°C
30°C (86°F)
6074-84-6
99.95%
Ta(OCH2CH3)5
MFCD00049785
3678999
145°C (0.1mmHg)
5g

Chemical Identifiers

6074-84-6
Ta(OCH2CH3)5
MFCD00049785
3678999
Tantalum(V) ethoxide
C10H25O5Ta
406.25
UN2924
Tantalum(V) ethylate
Optical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA