Learn More
Tantalum(V) ethoxide, 99.999% (metals basis), Nb <100ppm, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01464303
Description
Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tantalum(V) ethoxide | |
18°C to 19°C | |
145°C (0.1 mmHg) | |
(metals basis) | |
1.488 | |
MFCD00049785 | |
UN2924 | |
Air and moisture sensitive | |
Decomposes in water; Soluble in organic solvents. | |
CCO[Ta](OCC)(OCC)(OCC)OCC | |
406.25 | |
406.25 | |
Nb <100ppm |
6074-84-6 | |
1.566 g/mL | |
30°C (87°F) | |
C10H25O5Ta | |
Ta(OCH2CH3)5 | |
1 g | |
3678999 | |
tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide | |
HSXKFDGTKKAEHL-UHFFFAOYSA-N | |
ethanolate;tantalum(5+) | |
160806 | |
99.999% | |
Liquid |
Chemical Identifiers
6074-84-6 | |
406.25 | |
HSXKFDGTKKAEHL-UHFFFAOYSA-N | |
160806 | |
CCO[Ta](OCC)(OCC)(OCC)OCC |
C10H25O5Ta | |
MFCD00049785 | |
tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide | |
ethanolate;tantalum(5+) |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 228-010-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only