missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum(V) ethoxide, 99+%

Manufacturer:  Alfa Aesar™ L1028814

 View more versions of this product

Catalog No. AAL1028814



Description

Description

Applications
Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

Solubility
Soluble in organic solvents.

Notes
Moisture sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Tantalum(V) ethoxide
C10H25O5Ta
MFCD00049785
3678999
HSXKFDGTKKAEHL-UHFFFAOYSA-N
ethanolate;tantalum(5+)
160806
≥99%
18°C to 19°C
30°C (86°F)
6074-84-6
Ta(OCH2CH3)5
UN2924
tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Ta+5]
406.253
406.25
145°C (0.1mmHg)
25g

Chemical Identifiers

6074-84-6
406.253
HSXKFDGTKKAEHL-UHFFFAOYSA-N
160806
CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Ta+5]
C10H25O5Ta
MFCD00049785
tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
ethanolate;tantalum(5+)
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Documents