Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum(V) chloride, Puratronic™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Tantalum(V) chloride
CAS 7721-01-9
Purity Grade Puratronic™
Formula Weight 358.21
MDL Number MFCD00011253
UN Number UN3260
Merck Index 14,9054
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1470806

 
alfa aesar™
1470806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1470814

 
alfa aesar™
1470814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1470822

 
alfa aesar™
1470822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 358.21
Packaging Ampule
Chemical Name or Material Tantalum(V) chloride
TSCA TSCA
Mesh Size −60 Mesh
Odor Acidic
Purity Grade Puratronic™
Physical Form Resublimed Powder
Assay 99.99%
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis)
MDL Number MFCD00011253
CAS 7721-01-9
UN Number UN3260
Merck Index 14,9054
Boiling Point 242°C
Color White to pale yellow
Melting Point 216°C
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Density 3.68g/mL
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
RTECS Number WW5600000

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

  • Ampouled under argon