missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum(V) Chloride Anhydrous, TCI America™

Manufacturer:  TCI America T268425G

 View more versions of this product

Catalog No. T268425G



Specifications

Specifications

Tantalum(V) Chloride Anhydrous
White
25g
Cl5Ta
3260
358.20
Crystal-Powder at 20°C
239°C
220°C
7721-01-9
MFCD00011253
OEIMLTQPLAGXMX-UHFFFAOYSA-I
358.20
≥95.0% (T)

Chemical Identifiers

7721-01-9
358.20
OEIMLTQPLAGXMX-UHFFFAOYSA-I
Cl5Ta
MFCD00011253
Safety and Handling

Safety and Handling

EINECSNumber : (1)-0654

RTECSNumber : WW5600000

TSCA : Yes

Documents