missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum(V) Chloride, 99.99% (Metal Basis), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar 1088022

 View more versions of this product

Catalog No. AA1088022



Specifications

Specifications

Tantalum(V) Chloride
7721-01-9
Cl5Ta
OEIMLTQPLAGXMX-UHFFFAOYSA-I
358.21
Moisture sensitive
Ampouled under argon
99.99%, metal basis
MFCD00011253
358.20
Solid, Resublimed, Crystalline
100g

Chemical Identifiers

7721-01-9
358.20
OEIMLTQPLAGXMX-UHFFFAOYSA-I
Cl5Ta
MFCD00011253
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Harmful by inhalation, in contact with skin and if swallowed. Causes burns.

DOTInformation : DOT Class: 8

EINECSNumber : 231-755-6

Documents