Please login to your online account to display your discounted pricing

Tantalum(V) Chloride, 99.99% (Metal Basis), Alfa Aesar™, Puratronic™

Chemical Identifiers

Chemical Name or Material Tantalum(V) Chloride
Name Note Ampouled under argon
CAS 7721-01-9
Physical Form Solid, Resublimed, Crystalline
Assay Percent Range 99.99%, metal basis
Molecular Formula TaCl5
Formula Weight 358.21
MDL Number MFCD00011253
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1088006

  SDS
alfa aesar
1088006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1088014

  SDS
alfa aesar
1088014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1088022

  SDS
alfa aesar
1088022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tantalum(V) Chloride
Name Note Ampouled under argon
CAS 7721-01-9
Health Hazard 1 Harmful by inhalation, in contact with skin and if swallowed. Causes burns.
Health Hazard 2 When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
DOT Information DOT Class: 8
EINECS Number 231-755-6
Physical Form Solid, Resublimed, Crystalline
Assay Percent Range 99.99%, metal basis
Molecular Formula TaCl5
Formula Weight 358.21
MDL Number MFCD00011253
Sensitivity Moisture sensitive
Quantity 5g
Safety and Handling

Harmful by inhalation, in contact with skin and if swallowed. Causes burns.

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).