Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum(V) chloride, 99.8% (metals basis)

Chemical Identifiers

Chemical Name or Material Tantalum(V) chloride
CAS 7721-01-9
Formula Weight 358.21
MDL Number MFCD00011253
UN Number UN3260
Merck Index 14,9054
Synonym Tantalum pentachloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1416406

  SDS
alfa aesar™
1416406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1416414

  SDS
alfa aesar™
1416414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1416422

  SDS
alfa aesar™
1416422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 358.21
Packaging Ampule
Quantity 5g
Physical Form Powder
Merck Index 14,9054
Assay Percent Notes (metals basis)
Color White to pale yellow
Solubility Information Decomposes in water; Soluble in absolute alcohol and KOH
Boiling Point 242°C
Assay 99.8%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Tantalum pentachloride
RTECS Number WW5600000
Density 3.68g/mL
MDL Number MFCD00011253
Melting Point 216°C
Odor Acidic
CAS 7721-01-9
UN Number UN3260
Chemical Name or Material Tantalum(V) chloride
Sensitivity Moisture Sensitive
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA TSCA
Storage Note 1
  • Ampouled under argon

  • In chlorination of organic substances, as an intermediate, and in the production of pure metal
  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

  • Ampouled under argon