missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum(V) chloride, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 1416422

 View more versions of this product

Catalog No. AA1416422



Description

Description

  • In chlorination of organic substances, as an intermediate, and in the production of pure metal
Specifications

Specifications

Tantalum(V) chloride
(metals basis)
MFCD00011253
14,9054
Decomposes in water; Soluble in absolute alcohol and KOH
358.20
Powder
3.68g/mL
Ampule
242°C
216°C
7721-01-9
Cl5Ta
UN3260
Tantalum pentachloride
OEIMLTQPLAGXMX-UHFFFAOYSA-I
358.21
99.8%
Acidic
Moisture Sensitive
White to Yellow
100g

Chemical Identifiers

7721-01-9
358.20
OEIMLTQPLAGXMX-UHFFFAOYSA-I
Cl5Ta
MFCD00011253
Tantalum pentachloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : WW5600000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents