Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum(V) bromide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Tantalum(V) bromide
CAS 13451-11-1
Formula Weight 580.49
MDL Number MFCD00049563
UN Number UN3260
Synonym Tantalum pentabromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1404104

  SDS
alfa aesar™
1404104
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1404109

  SDS
alfa aesar™
1404109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1404118

  SDS
alfa aesar™
1404118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1404130

  SDS
alfa aesar™
1404130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 580.49
Chemical Name or Material Tantalum(V) bromide
TSCA TSCA
Mesh Size −8 Mesh
Odor Pungent
Physical Form Powder
Assay 99.9%
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis)
MDL Number MFCD00049563
CAS 13451-11-1
UN Number UN3260
Synonym Tantalum pentabromide
Boiling Point 348.8°C
Color Yellow
Melting Point 265°C
Quantity 2g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.67g/mL
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.