missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum tubing, 3.18mm (0.125in) OD, 2.26mm (0.089in) ID, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 44923BW

 View more versions of this product

Catalog No. AA44923BW



Specifications

Specifications

Tantalum tubing
OD: 3.18mm (0.125 in.), ID: 2.26mm (0.089 in.)
(metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
[Ta]
180.948
CHEBI:33348
50cm
7440-25-7
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
Odorless
tantalum
23956
99.9%

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents