missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum Thinfoil, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 14266GD

 View more versions of this product

Catalog No. AA14266GD



Specifications

Specifications

Tantalum Thinfoil
7440-25-7
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.948
CHEBI:33348
50 x 500mm
99.95% (metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
∽2.1g/100X100mm, Certified Pinhole Free

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

EINECSNumber : 231-135-5

Documents