Please login to your online account to display your discounted pricing

Tantalum standard solution, for AAS, 1mg/mL Ta in 5% HF, ACROS Organics™


Quantity 500mL
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  196335000

 View more versions of this product

Catalog No. AC196335000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.95
Physical Form Liquid
Quantity 500mL
Color Clear colorless
Purity Grade For AAS
CAS 7664-39-3
Molecular Formula Ta
CAS 7664-39-3
Concentration 995 to 1005 μg/mL of Ta in 5% HF (20°C)
Packaging Plastic bottle
CAS 7732-18-5
Health Hazard 1 GHS Signal Word: Danger
Name Note 1 mg/ml Ta in 5% HF
MDL Number MFCD00011252
Chemical Name or Material Tantalum standard solution
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tantalum standard solution
Name Note 1 mg/ml Ta in 5% HF
CAS 7664-39-3
CAS 7664-39-3
CAS 7732-18-5
Purity Grade For AAS
Molecular Formula Ta
Formula Weight 180.95
MDL Number MFCD00011252
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.

SDS & Safety Documents