Please login to your online account to display your discounted pricing

Tantalum standard solution, for AAS, 1mg/mL Ta in 5% HF, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tantalum standard solution
Name Note 1 mg/ml Ta in 5% HF
CAS 7664-39-3
CAS 7664-39-3
CAS 7732-18-5
Purity Grade For AAS
Molecular Formula Ta
Formula Weight 180.95
MDL Number MFCD00011252
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC196335000

  SDS
thermo fisher scientific
196335000
Plastic bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 180.95
Quantity 500mL
Color Clear colorless
Purity Grade For AAS
Name Note 1 mg/ml Ta in 5% HF
Molecular Formula Ta
Concentration 995 to 1005 μg/mL of Ta in 5% HF (20°C)
MDL Number MFCD00011252
CAS 7664-39-3
Health Hazard 1 GHS Signal Word: Danger
CAS 7664-39-3
Packaging Plastic bottle
CAS 7732-18-5
Chemical Name or Material Tantalum standard solution
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For atomic adsorption spectroscopy