missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum rod, 12.7mm (0.5 in.) dia., hard, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 10347BO

 View more versions of this product

Catalog No. AA10347BO



Specifications

Specifications

Tantalum rod
1cm
7440-25-7
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
Odorless
23956
Rod
≈20.4g/cm
12.7mm (0.5 in.) dia., Hard, Temper: as drawn
(metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
180.948
CHEBI:33348
99.95%

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents