missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tantalum powder, -100 mesh, Puratronic™, 99.98% (metals basis), Nb 50ppm, Thermo Scientific Chemicals
Nb 50ppm
Supplier: Thermo Scientific Chemicals 1034414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Black to Gray | |
7440-25-7 | |
16.69g/cm3 | |
Ta | |
25 g | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
tantalum | |
23956 | |
-100 Mesh | |
Powder |
Tantalum | |
Gray to black powder | |
99.98% (Metals basis) | |
MFCD00011252 | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
[Ta] | |
180.95 | |
CHEBI:33348 | |
Odorless |
Chemical Identifiers
7440-25-7 | |
180.95 | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
23956 | |
tantalum |
Ta | |
MFCD00011252 | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
CHEBI:33348 | |
[Ta] |
Safety and Handling
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : DOT Class: 4.1; Packaging Group: II
EINECSNumber : 231-135-5
RTECSNumber : WW5505000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only