missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum powder, 99.97% (metal basis), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar 1034522

 View more versions of this product

Catalog No. AA1034522



Specifications

Specifications

Tantalum
7440-25-7
16.69g/cm3
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
Odorless
23956
Powder
100g
Gray to black powder
99.97% (Metals basis, Nb 50ppm)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
180.948
CHEBI:33348
Black to Gray

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

H228-H315-H319-H335: Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-135-5

Documents