missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum Powder, -22 mesh, Puratronic™, 99.98% (Metals basis, Nb 50ppm), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1075322

 View more versions of this product

Catalog No. AA1075322



Specifications

Specifications

Tantalum
7440-25-7
99.98% (Metals basis, Nb 50ppm)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
180.948
CHEBI:33348
Black to Gray
100g
Gray to black powder
Ta
foil
[Ta]
Odorless
23956
Powder

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
foil
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

H228-H315-H319-H335; Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-135-5

Documents