missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum plate, 6.35mm (0.25 in.) thick, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 14181FI

 View more versions of this product

Catalog No. AA14181FI



Specifications

Specifications

Tantalum plate
50 x 50mm
7440-25-7
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
Odorless
23956
Plate
≈65.9g/25x25mm
6.35mm (0.25 in.) thick
(metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
180.948
CHEBI:33348
99.9%

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents