Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum, plasma standard solution, Specpure™, Ta 10,000μg/mL

Chemical Identifiers

Chemical Name or Material Tantalum plasma standard solution
Name Note Ta 10000μg/mL
Purity Grade Specpure™
MDL Number MFCD00011252
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14419AD

  SDS
alfa aesar™
14419AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14419AE

  SDS
alfa aesar™
14419AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14419AP

  SDS
alfa aesar™
14419AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Specpure™
Concentration TaCl5 in 2% HF
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P361-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Health Hazard 1 Danger
Name Note Ta 10000μg/mL
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H301-H311-H318-H315
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
UN Number UN3264
MDL Number MFCD00011252
Quantity 50mL
Chemical Name or Material Tantalum plasma standard solution
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P361-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H318-H315
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.