missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum, plasma standard solution, Specpure™, Ta 10,000μg/mL

Manufacturer:  Alfa Aesar™ 14419AP

 View more versions of this product

Catalog No. AA14419AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Tantalum plasma standard solution
MFCD00011252
TaCl5 in 2% HF
Specpure™
Ta 10000μg/mL
UN3264
Liquid
500mL

Chemical Identifiers

MFCD00011252
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H318-H315
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P361-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents