missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum nitride, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 1309309

 View more versions of this product

Catalog No. AA1309309



Specifications

Specifications

Tantalum nitride
(metals basis)
MFCD00049568
MZLGASXMSKOWSE-UHFFFAOYSA-N
azanylidynetantalum
82832
Powder
−325 Mesh
3360°C
12033-62-4
NTa
tantalum nitride, tantalum nitride tan, tantalum mononitride, tantalum nitride trace metals basis 10g
N#[Ta]
194.955
194.95
99.5%
Odorless
10g

Chemical Identifiers

12033-62-4
194.955
MZLGASXMSKOWSE-UHFFFAOYSA-N
82832
N#[Ta]
NTa
MFCD00049568
tantalum nitride, tantalum nitride tan, tantalum mononitride, tantalum nitride trace metals basis 10g
azanylidynetantalum
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents