Please login to your online account to display your discounted pricing

Tantalum ICP Standard, 1mL = 10mg Ta (10,000ppm Ta), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Tantalum ICP Standard
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 7664-39-3
CAS Min % 1.89
CAS Max % 1.96
CAS 12022-02-5
CAS Min % 1.83
CAS Max % 1.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PTA10KW-100

 
ricca chemical company
RIPTA10KW100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PTA10KW-50

 
ricca chemical company
RIPTA10KW50
Poly natural 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Tantalum ICP Standard
CAS Max % 1.9
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 7664-39-3
Packaging Poly natural
CAS Min % 1.89
CAS Max % 1.96
Concentration or Composition Notes 10,000ppm Ta
CAS 12022-02-5
Equivalence 1 mL = 10 mg Ta
CAS Min % 1.83