missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum gauze, 30 mesh woven from 0.076mm (0.003 in.) dia. wire

Manufacturer:  Alfa Aesar™ 43965GN

 View more versions of this product

Catalog No. AA43965GN



Specifications

Specifications

Tantalum gauze
30 mesh woven from 0.076mm (0.003 in.) dia. wire
Ta
30 Mesh
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
Wire Cloth
125 x 125mm
7440-25-7
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.948
CHEBI:33348

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Documents