missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 046695FL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tantalum gauze | |
7440-25-7 | |
MFCD00011252 | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
[Ta] | |
180.95 | |
CHEBI:33348 | |
Odorless |
100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave | |
Ta | |
75 x 75 mm | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
tantalum | |
23956 | |
100 Mesh | |
Wire Cloth |
Chemical Identifiers
7440-25-7 | |
180.95 | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
23956 | |
tantalum |
Ta | |
MFCD00011252 | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
CHEBI:33348 | |
[Ta] |
Safety and Handling
EINECSNumber : 231-13505
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only