missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum foil, 99.95% (metals basis), Alfa Aesar™

Form: approx. 6.3g/100x150mm; Temper: as rolled

Manufacturer:  Alfa Aesar 10351GJ

 View more versions of this product

Catalog No. AA10351GJ



Specifications

Specifications

Tantalum
Hard
Gray to silver foil
99.95% (Metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
180.948
CHEBI:33348
Gray to Silver
100 x 150mm
7440-25-7
16.69g/cm3
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
Odorless
23956
Foil, 0.025mm (0.001 in.) thick

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

EINECSNumber : 231-135-5

Documents