Please login to your online account to display your discounted pricing

Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tantalum foil
CAS 7440-25-7
Name Note 1.0mm (0.04 in.) thick
MDL Number MFCD00011252
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AA10356FF

  SDS
alfa aesar
10356FF
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density ≈10.4g/25x25mm
Chemical Name or Material Tantalum foil
Name Note 1.0mm (0.04 in.) thick
CAS 7440-25-7
MDL Number MFCD00011252
Odor Odorless
Assay 99.95%
TSCA TSCA
Assay Percent Notes (metals basis)