Please login to your online account to display your discounted pricing

Tantalum foil, 0.5mm (0.02in) thick, annealed, 99.95% (metals basis excluding Nb), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tantalum foil, 0.5mm (0.02 in.) thick, annealed
CAS 7440-25-7
MDL Number MFCD00011252
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AA10355GW

  SDS
alfa aesar
10355GW
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 99.95%
Odor Odorless
Chemical Name or Material Tantalum foil, 0.5mm (0.02 in.) thick, annealed
CAS 7440-25-7
MDL Number MFCD00011252
TSCA TSCA
Assay Percent Notes (metals basis excluding Nb)
Density ≈5.19g/25x25mm