Please login to your online account to display your discounted pricing

Tantalum foil, 0.127mm (0.005in) thick, annealed, 99.95% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tantalum foil
CAS 7440-25-7
Name Note 0.127mm (0.005 in.) thick, annealed
MDL Number MFCD00011252
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AA10352GW

  SDS
alfa aesar
10352GW
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Temper: Soft
MDL Number MFCD00011252
Assay Percent Notes (metals basis)
Assay 99.95%
CAS 7440-25-7
Density ≈5.3g/50x50mm
TSCA TSCA
Name Note 0.127mm (0.005 in.) thick, annealed
Odor Odorless
Chemical Name or Material Tantalum foil