missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum Foil, 0.127mm (0.005 in.) thick, annealed, 99.95% (Metals basis), Alfa Aesar™

Form: Approx. 5.3g/50 x 50mm; Temper: soft

Manufacturer:  Alfa Aesar 10352GH

 View more versions of this product

Catalog No. AA10352GH



Specifications

Specifications

Tantalum
Annealed
Gray to silver foil
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
Odorless
23956
Foil, 0.127mm (0.005 in.) thick
100 x 100mm
7440-25-7
99.95% (Metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
180.948
CHEBI:33348
Gray to Silver

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Documents