Please login to your online account to display your discounted pricing

Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tantalum foil
CAS 7440-25-7
Name Note 0.025mm (0.001 in.) thick, hard
MDL Number MFCD00011252
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AA10351IX

  SDS
alfa aesar
10351IX
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Temper: As Rolled
TSCA TSCA
Odor Odorless
Chemical Name or Material Tantalum foil
CAS 7440-25-7
Density ≈6.3g/100x150mm
Name Note 0.025mm (0.001 in.) thick, hard
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00011252