missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum disilicide

Manufacturer:  Alfa Aesar™ 4496518

 View more versions of this product

Catalog No. AA4496518



Specifications

Specifications

Tantalum disilicide
Si2Ta
tantalum disilicide, tantalum silicide tasi2, tantalum silicide, tasi2
[Si]=[Ta]=[Si]
237.118
237.12
−325 Mesh
50g
12039-79-1
MFCD00049564
MANYRMJQFFSZKJ-UHFFFAOYSA-N
bis($l^{2}-silanylidene)tantalum
6336888
Powder
2200°C

Chemical Identifiers

12039-79-1
237.118
MANYRMJQFFSZKJ-UHFFFAOYSA-N
6336888
[Si]=[Ta]=[Si]
Si2Ta
MFCD00049564
tantalum disilicide, tantalum silicide tasi2, tantalum silicide, tasi2
bis($l^{2}-silanylidene)tantalum
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents