Please login to your online account to display your discounted pricing

Alfa Aesar™ Tantalum carbide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Tantalum carbide
CAS 12070-06-3
Formula Weight 192.96
MDL Number MFCD00011255
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1214409

  SDS
alfa aesar™
1214409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1214418

  SDS
alfa aesar™
1214418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1214430

  SDS
alfa aesar™
1214430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.96
UN Number UN3178
Density 13.9g/mL
Odor Odorless
Quantity 10g
Chemical Name or Material Tantalum carbide
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Mesh Size −325 Mesh
Melting Point 3880°C
Physical Form Powder
CAS 12070-06-3
Color Gray to black
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Assay 99.5%
TSCA TSCA
Assay Percent Notes (metals basis)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
MDL Number MFCD00011255
Boiling Point 5500°C
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.