Please login to your online account to display your discounted pricing

Tanshinone IIA, Tocris™

Inhibits NF-κB and AP-1 DNA binding; Displays antioxidant properties

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material Tanshinone IIA
Assay Percent Range >98%
Molecular Formula C19H18O3
Formula Weight 294.34
Synonym 6,7,8,9-Tetrahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

442650

 
r & d systems
RDS442650
50mg Each for $924.23

442610

 
r & d systems
RDS442610
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 294.34
Quantity 50mg
Molecular Formula C19H18O3
Synonym 6,7,8,9-Tetrahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione
Chemical Name or Material Tanshinone IIA
Assay Percent Range >98%
CAS 568-72-9
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature