missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tanshinone IIA, TCI America™

Manufacturer:  TCI America T298750MG

 View more versions of this product

Catalog No. T298750MG



Specifications

Specifications

Tanshinone IIA
198°C
568-72-9
MFCD00238692
HYXITZLLTYIPOF-UHFFFAOYSA-N
1,6,6-trimethyl-8,9-dihydro-7H-naphtho[1,2-g][1]benzofuran-10,11-dione
164676
Crystal-Powder at 20°C
Undesignated
50mg
C19H18O3
tanshinone iia, tanshinone ii, tanshinone b, dan shen ketone, tanshinon ii, 1,6,6-trimethyl-6,7,8,9-tetrahydrophenanthro 1,2-b furan-10,11-dione, unii-4gpc9fqg6l, 4gpc9fqg6l, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g benzofuran-10,11-dione, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g 1 benzofuran-10,11-dione
CC1=COC2=C1C(=O)C(=O)C3=C2C=CC4=C3CCCC4(C)C
294.35
294.35
≥97.0% (HPLC)

Chemical Identifiers

568-72-9
294.35
HYXITZLLTYIPOF-UHFFFAOYSA-N
164676
CC1=COC2=C1C(=O)C(=O)C3=C2C=CC4=C3CCCC4(C)C
C19H18O3
MFCD00238692
tanshinone iia, tanshinone ii, tanshinone b, dan shen ketone, tanshinon ii, 1,6,6-trimethyl-6,7,8,9-tetrahydrophenanthro 1,2-b furan-10,11-dione, unii-4gpc9fqg6l, 4gpc9fqg6l, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g benzofuran-10,11-dione, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g 1 benzofuran-10,11-dione
1,6,6-trimethyl-8,9-dihydro-7H-naphtho[1,2-g][1]benzofuran-10,11-dione
Safety and Handling

Safety and Handling

RTECSNumber : SF8282695

TSCA : No

Recommended Storage : Refrigerator

Documents