missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tannic acid, ACS, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03641022

 View more versions of this product

Catalog No. AA3641022



Description

Description

Tannic acid has anti-bacterial, anti-enzymatic and astringent applications. Used in the dyeing process for cellulose fibers such as cotton. Used in food and medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tannic Acid
1401-55-4
MFCD00066397
tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
LRBQNJMCXXYXIU-JPZKJCTESA-N
1701.206
1701.23
100 g
Powder
∽218°C (decomposition)
C76H52O46
8186396
Soluble in water,warm glycerol,alcohol and acetone.
C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(
134129492
ACS Reagent
199°C (390°F)
Air and light sensitive

Chemical Identifiers

1401-55-4
1701.206
LRBQNJMCXXYXIU-JPZKJCTESA-N
134129492
C76H52O46
MFCD00066397
tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(
Safety and Handling

Safety and Handling

EINECSNumber : 215-753-2

RTECSNumber : WW5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only