Please login to your online account to display your discounted pricing

Tandospirone hydrochloride, Tocris

Selective 5-HT1A partial agonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material Tandospirone hydrochloride
Assay Percent Range >98%
Molecular Formula C21H29N5O2·HCl
Formula Weight 419.95
Synonym (3aR,4S,7R,7aS)-rel-Hexahydro-2-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-4,7-methano-1H-isoindole-1,3(2H)-dione hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

285450

 
r & d systems
285450
50mg Each for $762.58

285410

 
r & d systems
285410
10mg Each for $181.19
Description & Specifications

Specifications

Formula Weight 419.95
Quantity 50mg
Chemical Name or Material Tandospirone hydrochloride
Molecular Formula C21H29N5O2·HCl
Assay Percent Range >98%
CAS 99095-10-0
Synonym (3aR,4S,7R,7aS)-rel-Hexahydro-2-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-4,7-methano-1H-isoindole-1,3(2H)-dione hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C