Please login to your online account to display your discounted pricing

Tamsulosin hydrochloride, Tocris

Selective α1A antagonist

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material Tamsulosin hydrochloride
Assay Percent Range >99%
Molecular Formula C20H28N2O5S·HCl
Formula Weight 444.97
Synonym 5-[(2R)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-benzenesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

305010

 
r & d systems
RDS305010
10mg Each for $221.01

305050

 
r & d systems
RDS305050
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 444.97
Quantity 10mg
Molecular Formula C20H28N2O5S·HCl
CAS 106463-17-6
Assay Percent Range >99%
Synonym 5-[(2R)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-benzenesulfonamide hydrochloride
Chemical Name or Material Tamsulosin hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature