missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tamoxifen Citrate Salt, MP Biomedicals™
Manufacturer: Mp Biomedicals 0215673980
Description
Molecular biologySpecifications
Tamoxifen Citrate Salt | |
White | |
MFCD00058321 | |
100mg | |
OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 | |
563.65 | |
CHEBI:9397 | |
Solid |
54965-24-1 | |
C32H37NO8 | |
farmifeno, ginarsan, i.c.i. 46474 citrate, istubal, jenoxifen, tamoxifen citrate, tamoxifen citrate salt, unii-7frv7310n6, zitazonium | |
FQZYTYWMLGAPFJ-OQKDUQJOSA-N | |
(2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid | |
2733525 | |
563.65 |
Chemical Identifiers
54965-24-1 | |
563.65 | |
FQZYTYWMLGAPFJ-OQKDUQJOSA-N | |
2733525 | |
(2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid |
C32H37NO8 | |
MFCD00058321 | |
farmifeno, ginarsan, i.c.i. 46474 citrate, istubal, jenoxifen, tamoxifen citrate, tamoxifen citrate salt, unii-7frv7310n6, zitazonium | |
CHEBI:9397 | |
OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 259-415-2
storageNote1 : Store desiccated. Protect from light
Recommended Storage : Store at +4°C