Please login to your online account to display your discounted pricing

Taltirelin, Tocris

Synthetic thyrotropin-releasing hormone analog

$136.87 - $290.95

Chemical Identifiers

Chemical Name or Material Taltirelin
Assay Percent Range >96%
Molecular Formula C17H23N7O5
Formula Weight 405.41
Synonym N-[[(4S)-Hexahydro-1-methyl-2, 6-dioxo-4-pyrimidinyl]carbonyl]-L-histidyl-L-prolinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

26721

 
r & d systems
RDS26721
1mg Each for $136.87

267210

 
r & d systems
RDS267210
10mg Each for $290.95
Description & Specifications

Specifications

Formula Weight 405.41
Quantity 1mg
Chemical Name or Material Taltirelin
Assay Percent Range >96%
Molecular Formula C17H23N7O5
CAS 103300-74-9
Synonym N-[[(4S)-Hexahydro-1-methyl-2, 6-dioxo-4-pyrimidinyl]carbonyl]-L-histidyl-L-prolinamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C