missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Talc (Powder/USP), Fisher Chemical

Manufacturer:  Thermo Scientific FLT2212

 View more versions of this product

Catalog No. T2-212



Specifications

Specifications

Talc
14807-96-6,14808-60-7
2.0% max. (USP)
MFCD00084820
Powder/Solid
0.9% max. (USP)
0.25% max. (USP)
7.0% max. (USP)
Poly Pail
0.1% max. (USP)
White
2.5kg
Pass Test (USP)
Pass Test (USP)
3MgO4SiO2H2O
Soapstone, Talcum
USP
Pass Test (USP)
0.001% max. (USP)
17.0 to 19.5% (USP)
Meets Requirements (USP)
5.7g/mol
800°C
Documents