Please login to your online account to display your discounted pricing

Talc, Powder, Multi-Compendial, J.T.Baker™

$178.90 - $204.62

Chemical Identifiers

Chemical Name or Material Talc
CAS 14807-96-6
Purity Grade Multi-Compendial
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004446

 
avantor performance
410005
Poly Bottle 2kg Each for $204.62

02004447

 
avantor performance
410001
Poly Bottle 500g Each for $178.90
Description & Specifications

Specifications

Physical Form Powder
Chemical Name or Material Talc
CAS 14807-96-6
Packaging Poly Bottle
Purity Grade Multi-Compendial
Quantity 2kg